missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPMT Antibody (OTI2A2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74570H
This item is not returnable.
View return policy
Description
TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TPMT | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TPMT | |
| Full length recombinant protein of human TPMT (NP_000358) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2A2 | |
| Western Blot, Immunohistochemistry | |
| EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes, Signal Transduction | |
| 7172.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction