Learn More
Ebro Electronic TPN351/EB14-A-NSMP NiCr-Ni Surface Probe 15mm 500C max SMP connector
Brand:  Ebro Electronic 1343-0712
TPN351/EB14-A-NSMP NiCr-Ni Surface Probe 15mm 500C max SMP connector
Product Code. 18536897
  • ONLINE EXCLUSIVE PRICE
    € 595.00
    € 492.00 / Each
    Save € 103.00
    17% Off
Only null left
Estimated Shipment: 29-01-2026