missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP/p25 Antibody (SR1385), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22044-50ul
This item is not returnable.
View return policy
Description
TPPP/p25 Monoclonal antibody specifically detects TPPP/p25 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| TPPP/p25 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| SR1385 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| p24, P24,25 kDa brain-specific protein, P25, p25alpha, p25-alpha, TPPP/p25brain specific protein p25 alpha, TPPP1glycogen synthase kinase 3 (GSK3) inhibitor p24, tubulin polymerization promoting protein, tubulin polymerization-promoting protein | |
| A synthesized peptide derived from human TPPP/p25 (Uniprot #: O94811) | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 11076 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction