missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97003H
This item is not returnable.
View return policy
Description
TPPP2 Polyclonal specifically detects TPPP2 in Human samples. It is validated for ELISA.
Specifications
| TPPP2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 122664 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| HRP | |
| C14orf8, chromosome 14 open reading frame 8, P18, p25beta, Protein p25-beta, TPPP/p18, tubulin polymerization-promoting protein family member 2 | |
| Produced in rabbits immunized with purified, recombinant TPPP2 (AAH38970.1; Met1-Lys170). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction