missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00282APC
This item is not returnable.
View return policy
Description
TPPP3 Polyclonal antibody specifically detects TPPP3 in Human samples. It is validated for ELISA
Specifications
| TPPP3 | |
| Polyclonal | |
| ELISA | |
| brain specific protein, CGI-38, p20, p25gamma, TPPP/p20, tubulin polymerization-promoting protein family member 3 | |
| Produced in rabbits immunized with purified, recombinant Human TPPP3 (Uniprot#: Q9BW30; Met1-Lys176) | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 51673 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction