missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPRKB Antibody (OTI3H3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74574APC
This item is not returnable.
View return policy
Description
TPRKB Monoclonal specifically detects TPRKB in Human, Mouse, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications
| TPRKB | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| TPRKB | |
| Full length recombinant protein of human TPRKB (NP_057142) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Monkey, Primate | |
| IgG1 |
| Flow Cytometry | |
| OTI3H3 | |
| Flow Cytometry | |
| CGI-121, PRPK (p53-related protein kinase)-binding protein, PRPK-binding protein, TP53RK binding protein, TP53RK-binding protein | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 51002.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction