missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPSB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 213.00 - € 486.00
Specifications
| Antigen | TPSB2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17740884
![]() |
Invitrogen™
PA5119480 |
200 μL |
€ 486.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17702934
|
Invitrogen™
PA5121532 |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TPSB2 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
TPSB2 is a protein coding gene. Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. These genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. The alleles of this gene exhibit an unusual amount of sequence variation, such that the alleles were once thought to represent two separate genes, beta II and beta III. Beta tryptases appear to be the main isoenzymes expressed in mast cells, whereas in basophils, alpha-tryptases predominate. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders.Specifications
| TPSB2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Tpsb2 | |
| AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C | |
| Tpsb2 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P21845 | |
| 17229 | |
| Recombinant protein Tryptase Beta 2. The antigen corresponds to amino acid range 30-276 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title