missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPSG1 Monoclonal Antibody (OTI3F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504030S
This item is not returnable.
View return policy
Description
TPSG1 Monoclonal antibody specifically detects TPSG1 in Human samples. It is validated for Western BlotSpecifications
| TPSG1 | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRR2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25823 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3F2 | |
| Unconjugated | |
| TPSG1 | |
| PRSS31; serine protease 31; TMT; TPSG1; Transmembrane tryptase; trpA; Tryptase gamma; Tryptase- gamma; tryptase gamma 1; Tryptase gamma heavy chain; tryptase gamma I; tryptase gamma II; Tryptase gamma light chain | |
| Human recombinant protein fragment corresponding to amino acids 20-283 of human TPSG1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction