missing translation for 'onlineSavingsMsg'
Learn More

TPT-260 Dihydrochloride, MedChemExpress

Product Code. 16438507
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438507

Brand: Medchem Express HY13769A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TPT-260 Dihydrochloride (NSC55712) is a thiophene thiourea derivative with molecule weight 260.00 in free base form; There is no formal name yet, we temporally call this molecule as TPT-260.

TPT-260 Dihydrochloride (NSC55712) is a thiophene thiourea derivative with molecule weight 260.00 in free base form

Specifications

Chemical Name or Material TPT-260 Dihydrochloride
Physical Form Solid
CAS 2076-91-7
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H14Cl2N4S3
Synonym NSC55712 TPU-260 Dihydrochloride
Solubility Information DMSO : 100 mg/mL (300.01 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (99.99 mM; Need ultrasonic)
SMILES NC(SCC1=CC=C(CSC(N)=N)S1)=N.Cl.Cl
Molecular Weight (g/mol) 333.32
Formula Weight 333.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.