missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13422387
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13422387 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13422387 Supplier Invitrogen™ Supplier No. PA528273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (95%), Rat (95%), Pig (100%), Rabbit (98%), Chicken (90%), Sheep (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%), Guinea pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TPT1 (translationally controlled tumor protein 1) is a 23 kDa member of the TCTP family of calcium binding proteins. TPT1 is localized in the cytoplasm and widely expressed and serves as a transcriptional activator, calcium transporter, histamine inducer and antiapoptotic caspase 3 inhibitor. TPT1 is a cytokine-like molecule that causes the release of histamine, IL-4 and IL-13 from basophils as well as the secretion of IL-8 and a calcium response in eosinophils. TPT1 plays a pivotal role in allergic diseases and due to its wide distribution in brain, is thought to be involved in neurodegenerative disorders, such as Alzheimer's disease and Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPT1
Applications Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TPT1
Gene Accession No. P13693, P63028
Gene Alias 21 kDa polypeptide; 21 kDa polypeptide under translational control; fortilin; Histamine-releasing factor; HRF; Lens epithelial protein; p02; p21; p23; TCTP; Tpt1; translationally regulated transcript (21 kDa); translationally-controlled tumor protein; Trt; tumor protein, translationally-controlled 1
Gene Symbols TPT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TCTP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22070, 7178
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.