missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPX2 Antibody (18D5-1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-74556-0.025mg
This item is not returnable.
View return policy
Description
TPX2 Monoclonal specifically detects TPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TPX2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 0.5 - 1.0 μg/mL | |
| C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| TPX2 (18D5-1) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 18D5-1 | |
| Unconjugated | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| TPX2 | |
| Recombinant human TPX2 | |
| 0.025 mg | |
| Cell Cycle and Replication, Mitotic Regulators | |
| 22974 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction