missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPX2 Antibody (TPX2-01), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-67265MFV450
This item is not returnable.
View return policy
Description
TPX2 Monoclonal antibody specifically detects TPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| TPX2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| TPX2-01 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunocytochemistry | |
| C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis) | |
| Human TPX2 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators | |
| 22974 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction