Learn More
IKA™ TR 4 Tube Insert
Brand: IKA™ 0004525400
Additional Details : Weight : 0.10000kg
Specifications
TR 4 Tube Insert | |
IKA Trayster Basic and Trayster Digital, Suitable 6 x 50mL Tubes with 28mm dia. | |
1 Ea. |