missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 218.00 - € 519.00
Specifications
| Antigen | TR alpha/NR1A1/Thyroid Hormone Receptor alpha |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18408311
|
Novus Biologicals
NBP2-22523-50ug |
50 ug |
€ 218.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18127441
|
Novus Biologicals
NBP2-22523 |
200 μg |
€ 519.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Polyclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TR alpha/NR1A1/Thyroid Hormone Receptor alpha | |
| Western Blot 1:100, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7 | |
| THRA | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 1 mg/ml BSA. with 0.05% Sodium Azide | |
| 7067 | |
| Synthetic Peptide: S(455) R A V C G E D D S S E A S S(469) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title