missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR2/NR2C1 Antibody (OTI4C7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74576B
This item is not returnable.
View return policy
Description
TR2/NR2C1 Monoclonal specifically detects TR2/NR2C1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TR2/NR2C1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| NR2C1 | |
| Recombinant protein fragment corresponding to amino acids 1-237 of human TR2/NR2C1 (NP_001027458) produced in E.coli. | |
| 0.1 mL | |
| 7181.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4C7 | |
| Western Blot | |
| nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction