missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR4/NR2C2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97138F
This item is not returnable.
View return policy
Description
TR4/NR2C2 Polyclonal specifically detects TR4/NR2C2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TR4/NR2C2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor | |
| Produced in rabbits immunized with E. coli-derived Human TR4/NR2C2 fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 7182 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction