missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA-1-85/CD147 Antibody (TRA-1-85) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3195X
This item is not returnable.
View return policy
Description
TRA-1-85/CD147 Monoclonal antibody specifically detects TRA-1-85/CD147 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out
Specifications
| TRA-1-85/CD147 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| 2120Ep human embryonal carcinoma cell line | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out | |
| TRA-1-85 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction