missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA16 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | TRA16 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678941
|
Novus Biologicals
NBP2-95151-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686952
|
Novus Biologicals
NBP2-95151-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRA16 Polyclonal antibody specifically detects TRA16 in Human samples. It is validated for Western BlotSpecifications
| TRA16 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| nuclear receptor 2C2-associated protein, repressor for TR4 transactivation, TR4 orphan receptor associated protein TRA16, TRA16TR4 orphan receptor-associated 16 kDa protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-139 of human NR2C2AP (NP_795361.1). MTHSLVCPETVSRVSSVLNRNTRQFGKKHLFDQDEETCWNSDQGPSQWVTLEFPQLIRVSQLQIQFQGGFSSRRGCLEGSQGTQALHKIVDFYPEDNNSLQTFPIPAAEVDRLKVTFEDATDFFGRVVIYHLRVLGEKV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 126382 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts