Learn More
Invitrogen™ Trabikibart Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558864
 View more versions of this product
Product Code. 30283296
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Trabikibart, also known as CSL311, is a fully human monoclonal antibody that targets the common beta chain (beta c) receptor, shared by the interleukin-3 (IL-3), granulocyte-macrophage colony-stimulating factor (GM-CSF), and interleukin-5 (IL-5) cytokines. Colony stimulating factor 2 receptor subunit beta (CSF2RB), also known as CD131, is a critical component of the cytokine receptor family involved in hematopoiesis and immune response regulation. Located on chromosome 22q13.1, the CSF2RB gene encodes the common beta-subunit that pairs with the alpha-subunits of receptors for granulocyte-macrophage colony-stimulating factor (GM-CSF), interleukin-3 (IL-3), and interleukin-5 (IL-5). Structurally, the beta-subunit contributes to the formation of high-affinity receptor complexes essential for signal transduction. Functionally, upon ligand binding, CSF2RB stimulates various intracellular signaling cascades that mediate cellular proliferation, differentiation, and survival, particularly of hematopoietic progenitor cells and immune cells. Aberrations in CSF2RB expression or function can lead to immune dysfunctions and are associated with diseases such as asthma and leukemia. Recent studies have highlighted the importance of the CSF2RB in regulatory T cells, underscoring its role in autoimmune disease prevention.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Trabikibart
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG4SP