missing translation for 'onlineSavingsMsg'
Learn More

Trace Amine Receptor 1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 201.00 - € 481.00

Specifications

Antigen Trace Amine Receptor 1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18653221
View Documents
Novus Biologicals
NBP2-93938-0.02ml
0.02 mL
€ 201.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643531
Unterlagen ansehen
Novus Biologicals
NBP2-93938-0.1ml
0.1 mL
€ 481.00
0.10mL
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trace Amine Receptor 1 Polyclonal antibody specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Trace Amine Receptor 1
Western Blot
Unconjugated
Rabbit
GPCR
PBS (pH 7.3), 50% glycerol
134864
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAAR1 (NP_612200.1). TSTDIMLSSASIFHLSFISIDRYYAVCDPLRYKAKMNILVICVMIFISWSVPAVFAFGMIFLELNFKGAEEIYYKHVHCRGGCSVFFSKISGVLTFMTSFY
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.