Learn More
Thermo Scientific™ TRACE™ TR-225 GC Columns
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
15 m
30 m
60 m
Unit Size:
Each
Description
Thermo Scientific™ TRACE TR-225 GC Columns offer reliable and reproducible performance.
- Mid-polarity phase, 50% Cyanopropylphenyl polysiloxane
- Low bleed even at elevated temperatures
- Outstanding robustness for difficult separations
- Manufactured to minimize risk of damage from contaminated carrier gas
- Similar to: DB-225, HP-225, RTX-225, BP225
Recommended for:
- Fatty Acid Methyl Esters (FAMEs)
- Carbohydrates
- Neutrol Sterols
Specifications
Specifications
| Max. Temperature | 230/250°C |
| Diameter (Metric) | 0.25 mm |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Trace™ TR-225 |
| For Use With (Application) | GC for Fatty Acid Methyl Esters (FAMEs), GC for Carbohydrates, GC for Neutral Sterols |
| Film Thickness | 0.25 μm |
| Length (Metric) | 60 m |
| Polarity | Mid-Polar |