missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Silicium Standard for ICP, 10,000 mg/L Si in Nitric Acid, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.Certified content incl. uncertainty and expiry date are given on the certificate.
Specifications
Specifications
| CAS | 16961-83-4 |
| Product Line | TraceCERT |
| Purity Grade Notes | 10,000 mg/L Si in 5% nitric acid (contains HF traces), prepared with high purity Si, HNO3, HF and water |
| Concentration | 10,000 mg/L Si in nitric acid |
| Type | Certified Reference Material |
| Quantity | 100 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction