missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRACP/PAP/ACP5 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41178JF549
This item is not returnable.
View return policy
Description
TRACP/PAP/ACP5 Polyclonal antibody specifically detects TRACP/PAP/ACP5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TRACP/PAP/ACP5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Janelia Fluor 549 | |
| HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human TRAP. The immunogen is located within the last 50 amino acids of TRAP. Amino Acid Squence: EASGKSLFKTRLPRRARP | |
| 0.1 mL | |
| Protein Phosphatase | |
| 54 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction