missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRACP/PAP/ACP5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41178MFV610
This item is not returnable.
View return policy
Description
TRACP/PAP/ACP5 Polyclonal antibody specifically detects TRACP/PAP/ACP5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| TRACP/PAP/ACP5 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human TRAP. The immunogen is located within the last 50 amino acids of TRAP. Amino Acid Squence: EASGKSLFKTRLPRRARP | |
| 0.1 mL | |
| Protein Phosphatase | |
| 54 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction