missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRACP/PAP/ACP5 Antibody (rACP5/1070), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75780CL1
This item is not returnable.
View return policy
Description
TRACP/PAP/ACP5 Monoclonal antibody specifically detects TRACP/PAP/ACP5 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Protein Array
Specifications
| TRACP/PAP/ACP5 | |
| Monoclonal | |
| CoraFluor 1 | |
| HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
| Recombinant full-length human TRACP/PAP/ACP5 protein (Uniprot: P13686) | |
| 0.1 mL | |
| Protein Phosphatase | |
| 54 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| rACP5/1070 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction