missing translation for 'onlineSavingsMsg'
Learn More

TRACP/PAP/ACP5 Antibody (SPM601), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-11536AF647

Product Code. 18528674

  • € 774.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRACP/PAP/ACP5 Monoclonal antibody specifically detects TRACP/PAP/ACP5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRACP/PAP/ACP5
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat
Purified
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SPM601
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
Recombinant full-length human TRACP/PAP/ACP5 protein (Uniprot: P13686)
0.1 mL
Protein Phosphatase
54
Store at 4°C in the dark.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.