missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-1 Antibody (TRAF1/3365), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-27024CL1
This item is not returnable.
View return policy
Description
TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for ELISA
Specifications
| TRAF-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1 | |
| Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research | |
| 7185 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| TRAF1/3365 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction