missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody (5C2C3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 210.00 - € 552.00
Specifications
| Antigen | TRAF-2 |
|---|---|
| Clone | 5C2C3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271773
|
Novus Biologicals
NBP2-61661 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624218
|
Novus Biologicals
NBP2-61661-0.025ml |
0.025 mL |
€ 210.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF-2 Monoclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TRAF-2 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| 7186 | |
| Purified recombinant fragment of human TRAF2 (AA: 39-188) expressed in E. Coli. | |
| Primary | |
| 55.8 kDa |
| 5C2C3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| TRAF2 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title