missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-3 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76642DL594
This item is not returnable.
View return policy
Description
TRAF-3 Polyclonal antibody specifically detects TRAF-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TRAF-3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human. The immunogen is located within amino acids 20 - 70 of TRAF3. Amino Acid Squence: GYKEKFVKTVEDKYK | |
| 0.1 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 7187 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction