missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-4 Antibody (SR1969), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21648-100ul
This item is not returnable.
View return policy
Description
TRAF-4 Monoclonal antibody specifically detects TRAF-4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| TRAF-4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1969 | |
| Western Blot 1:1000-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A | |
| A synthesized peptide derived from human TRAF-4 (Uniprot #: Q9BUZ4) | |
| 100 μg | |
| Cancer, Zinc Finger | |
| 9618 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction