missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-5 Antibody (55A219) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB600-1107CL1
This item is not returnable.
View return policy
Description
TRAF-5 Monoclonal antibody specifically detects TRAF-5 in Human samples. It is validated for Western Blot
Specifications
| TRAF-5 | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5 | |
| Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction, Zinc Finger | |
| 7188 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot | |
| 55A219 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction