Learn More
TRAF-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-56436SS
This item is not returnable.
View return policy
Description
TRAF-6 Polyclonal specifically detects TRAF-6 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TRAF-6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.02% Sodium Azide | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1.0 mg/mL | |
| Western Blot 3-5 ug/ml, Flow Cytometry | |
| Q9Y4K3 | |
| TRAF6 | |
| Anti-TRAF6 polyclonal antibody was raised against a peptide corresponding to amino acids between 410 and 460 of human TRAF6. | |
| 0.025 mg | |
| Immunology, Innate Immunity, Signal Transduction, Zinc Finger | |
| 7189 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |