missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAF2 Recombinant Rabbit Monoclonal Antibody (3S2A8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551192
This item is not returnable.
View return policy
Description
Immunogen sequence: DGCGKKKIPR EKFQDHVKTC GKCRVPCRFH AIGCLETVEG EKQQEHEVQW LREHLAMLLS SVLEAKPLLG DQSHAGSELL QRCESLEKKT ATFENIVCVL NREVERVAMT AEACSRQHRL DQDKIEALSS KVQQLERSIG LKDLAMADLE QKVLEMEAST Y.
Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications
| TRAF2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P39429, Q12933 | |
| TRAF2 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 3S2A8 | |
| Unconjugated | |
| TRAF2 | |
| AI325259; E3 ubiquitin-protein ligase TRAF2; MGC:45012; OTTHUMP00000064745; RING-type E3 ubiquitin transferase TRAF2; TNF receptor associated factor 2; Tnf receptor-associated factor 2; TRAF2; TRAP; TRAP3; tumor necrosis factor type 2 receptor associated protein 3; tumor necrosis factor type 2 receptor-associated protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22030, 311786, 7186 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction