missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97341R
This item is not returnable.
View return policy
Description
TRAF3IP1 Polyclonal antibody specifically detects TRAF3IP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRAF3IP1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| DKFZp434F124, Interleukin-13 receptor alpha 1-binding protein 1, Microtubule-interacting protein associated with TRAF3, microtubule-interacting protein that associates with TRAF3, MIPT3interleukin 13 receptor alpha 1-binding protein-1, MIP-T3microtubule interacting protein that associates with TRAF3, TNF receptor-associated factor 3 interacting protein 1, TRAF3-interacting protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human TRAF3IP1 fragment. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction | |
| 26146 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction