missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32704
This item is not returnable.
View return policy
Description
TRAF3IP1 Polyclonal specifically detects TRAF3IP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAF3IP1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp434F124, Interleukin-13 receptor alpha 1-binding protein 1, Microtubule-interacting protein associated with TRAF3, microtubule-interacting protein that associates with TRAF3, MIPT3interleukin 13 receptor alpha 1-binding protein-1, MIP-T3microtubule interacting protein that associates with TRAF3, TNF receptor-associated factor 3 interacting protein 1, TRAF3-interacting protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26146 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TRAF3IP1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IGKCCLNKLSSDDAVRRVLAGEKGEVKGRASLTSRSQELDNKNVREEESRVHKNTEDRGDAEIKERSTSRDRKQKEELKEDR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction