missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | TRAF3IP1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633352
|
Novus Biologicals
NBP2-97341-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601645
|
Novus Biologicals
NBP2-97341-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF3IP1 Polyclonal antibody specifically detects TRAF3IP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TRAF3IP1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction | |
| PBS (pH 7.0) | |
| 26146 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp434F124, Interleukin-13 receptor alpha 1-binding protein 1, Microtubule-interacting protein associated with TRAF3, microtubule-interacting protein that associates with TRAF3, MIPT3interleukin 13 receptor alpha 1-binding protein-1, MIP-T3microtubule interacting protein that associates with TRAF3, TNF receptor-associated factor 3 interacting protein 1, TRAF3-interacting protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human TRAF3IP1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title