Learn More
TRAF3IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14404-1-AP-150UL
 View more versions of this product
Product Code. 16863024
  • € 410.00 / 150µL
Description
Tumor necrosis factor receptor (TNFR) superfamily members transmit signals regulating proliferation, differentiation and apoptosis in various types of cells. TNFR-associated factors (TRAFs) are a family of proteins that were initially discovered as downstream signal transducers of the TNFR superfamily. TRAF3 contains an N-terminal ring finger/zinc finger region that is thought to be essential for downstream signaling. MIP-T3 is associated with TRAF3. MIP-T3 binds to taxol-stabilized microtubules and to tubulin in vitro, and MIP-T3 recruits TRAF3 to microtubules when both proteins are overexpressed. The MIP-T3/TRAF3 interaction requires the coiled-coil TRAF-N domain of TRAF3. This interaction may provide a novel mechanism in sequestering TRAF3 to the cytoskeletal network.
Specifications
Specifications
TRAF3IP1
Polyclonal
Unconjugated
TRAF3IP1
MIP T3, MIPT3, TRAF3 interacting protein 1, TRAF3IP1
Rabbit
Antigen Affinity Chromatography
RUO
26146, 363286, 74019
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q149C2, Q5XIN3, Q8TDR0
Traf3ip1
TRAF3IP1 Fusion Protein Ag5677
150 μL
Primary
Human, Mouse, Rat