missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93556MFV610
This item is not returnable.
View return policy
Description
TRAF3IP2 Polyclonal antibody specifically detects TRAF3IP2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRAF3IP2 | |
| Polyclonal | |
| Western Blot | |
| ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2 | |
| Synthetic peptide corresponding to the N-terminus of human ACT. | |
| 0.1 mL | |
| Signal Transduction | |
| 10758 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction