missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF5 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810290S
This item is not returnable.
View return policy
Description
TRAF5 Monoclonal antibody specifically detects TRAF5 in Human samples. It is validated for Western BlotSpecifications
| TRAF5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NR83 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56731 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G3 | |
| Unconjugated | |
| SLC2A4RG | |
| 2700049M22Rik; AI875481; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; MGC:39780; p11 BID; p13 BID; p15 BID; p22 BID; RING finger protein 84; RNF84; TNF receptor associated factor 5; TNF receptor-associated factor 5; Traf5 | |
| Human recombinant protein fragment corresponding to amino acids 1-259 of human SLC2A4RG produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction