Learn More
Invitrogen™ TRAFD1 Monoclonal Antibody (8E6E7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517190
Description
MA5-17190 targets TRAFD1 in indirect ELISA, FACS, ICC, IF, and WB applications and shows reactivity with Human samples. The MA5-17190 immunogen is purified recombinant fragment of human TRAFD1 (amino acids: 401-582) expressed in E. Coli. MA5-17190 detects TRAFD1 which has a predicted molecular weight of approximately 64.8kDa.
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
Specifications
| TRAFD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O14545 | |
| TRAFD1 | |
| Purified recombinant fragment of human TRAFD1 (amino acids: 401-582) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 8E6E7 | |
| Unconjugated | |
| TRAFD1 | |
| 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10906 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.