missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAFD1 Monoclonal Antibody (OTI3E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809366
This item is not returnable.
View return policy
Description
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.Specifications
| TRAFD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14545 | |
| TRAFD1 | |
| Human recombit protein fragment corresponding to amino acids 308-582 of human TRAFD1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3E8 | |
| Unconjugated | |
| TRAFD1 | |
| FLN29 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10906 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction