missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PVEESIIIPC EFCGVQLEEE VLFHHQDQCD QRPATATNHV TEGIPRLDSQ PQETSPELPR RRVRHQGDLS SGYLDDTKQE TANGPTSCLP PSRPINNMTA TYNQLSRS Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 69%.
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
Specifications
Specifications
| Antigen | TRAFD1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRAFD1 |
| Gene Accession No. | O14545 |
| Gene Alias | 1110008K06Rik; Fln29; FLN29 gene product; Protein FLN29; TRAF type zinc finger domain containing 1; TRAFD1; TRAF-type zinc finger domain containing 1; TRAF-type zinc finger domain-containing protein 1 |
| Gene Symbols | TRAFD1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
