missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL/TNFSF10 Antibody (55B709.3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56518F
This item is not returnable.
View return policy
Description
TRAIL/TNFSF10 Monoclonal specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAIL/TNFSF10 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TNFSF10 | |
| This monoclonal antibody was raised against a peptide corresponding to amino acids 17-35 of human TRAIL. This peptide sequence has been shown to be involved in TRAIL and DR5 (TRAIL-R2) interaction. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors | |
| 8743 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 55B709.3 | |
| Immunohistochemistry-Paraffin | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In Jurkat, a 32 kDa band, which represents a membrane bound form of TRAIL, is observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction