missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL/TNFSF10 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-220JF646
This item is not returnable.
View return policy
Description
TRAIL/TNFSF10 Polyclonal antibody specifically detects TRAIL/TNFSF10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRAIL/TNFSF10 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors | |
| 8743 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction