missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (32A242), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56528FR
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| TRAILR1/TNFRSF10A | |
| Monoclonal | |
| DyLight 680 | |
| O00220 | |
| TNFRSF10A | |
| This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 8797 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 32A242 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, a 57 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction