missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR2/TNFRSF10B Antibody (B-D37), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14579AF405
This item is not returnable.
View return policy
Description
TRAILR2/TNFRSF10B Monoclonal antibody specifically detects TRAILR2/TNFRSF10B in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| TRAILR2/TNFRSF10B | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
| Recombinant human TRAILR2/TNFRSF10B | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway | |
| 8795 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA | |
| B-D37 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction