missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRAILR2/TNFRSF10B Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 8795 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56618 |
| Gene Alias | CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2 L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2 B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10 B, tumor necrosis factor receptor superfamily, member 10 b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 |
| Gene Symbol | TNFRSF10B |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction