missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TRAILR2/TNFRSF10B Antibody (OTI2A5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74578AF750
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TRAILR2/TNFRSF10B Monoclonal specifically detects TRAILR2/TNFRSF10B in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| TRAILR2/TNFRSF10B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TNFRSF10B | |
| Recombinant protein fragment corresponding to amino acids 203-411 of human TRAIL R2/TNFRSF10B (NP_671716) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2A5 | |
| Western Blot, Immunohistochemistry | |
| CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Biologically Active Proteins, Cancer, Death Receptor Signaling Pathway | |
| 8795.0 | |
| Store at 4°C in the dark. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering