missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Antibody (104918), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB633F
This item is not returnable.
View return policy
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal antibody specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), CyTOF
Specifications
| TRAILR4/TNFRSF10D/DcR2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 104918 | |
| Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready | |
| CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
| Mouse myeloma cell line NS0-derived recombinant human TRAIL R4/TNFRSF10D, Ala56-His211, Accession # Q9UBN6 | |
| 0.5 mL | |
| Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 8793 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction