missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIP Antibody (OTI2D4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74580B
This item is not returnable.
View return policy
Description
TRAIP Monoclonal specifically detects TRAIP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| TRAIP | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TRAIP | |
| Recombinant protein fragment corresponding to amino acids 67-313 of human TRAIP (NP_005870) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2D4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 10293.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction